Learn More
Creative Biomart Recombinant Giardia lamblia Antigen, His-tagged
Supplier:  Creative Biomart H3N2309
Encompass_Preferred
The E. coli derived recombinant Giardia intestinalis protein contains 266 amino acids (21-280). Giardia lamblia protein is fused to 6 His tag at C-terminal and purified by proprietary chromatograph
Catalog No. 50-208-4687
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.