Learn More
Sino Biological Influenza A H3N2 (A/Darwin/9/2021) Hemagglutinin / HA trimer Protein (His Tag)(MALS-verified)
Supplier:  Sino Biological 40859V08H1100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Darwin/9/2021) (H3N2)) hemagglutinin (translated amino acids of EPI1859996), termed as HA, was expressed with the bacteriophage T4 fibritin and a polyhistidine tag at the C-terminus.
Catalog No. 50-240-9774