Learn More
Sino Biological H3N2 NA Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40869V08B1100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Darwin/6/2021) Neuraminidase (EPI1925254) (Ile30-Ile469) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2609
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.