Learn More
Creative Biomart Meningococcal polysaccharide Group A Vaccine Antigen
Supplier:  Creative Biomart H3N2317
Encompass_Preferred
Meningococcal group A polysaccharide is group-specific polysaccharide antigens extracted and purified from Neisseria meningitidis serogroup A. N. meningitidis is a bacteria that causes endemic and epi
Catalog No. 50-208-4695
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.