Learn More
Alkali Scientific Hermle Z446-K Universal Centri
Supplier: Alkali Scientific RS6210
The Hermle Z446 universal High Capacity centrifuge series offers the highest capacity of any benchtop Hermle-Alkali model With air-craft grade premium alloy construction the 4 x 750ml swing out rotor can withstand centrifugal forces upwards of 4500 xg with full loads of up to 28 x 50ml 68 x 15ml tubes and up to 20 x microplates Advanced microprocessor controls allow for instant rotor identificationwhich automatically converts the rotational speed to the relevant g-force (rcf) preventing the user from setting a speed above the capabilities of the installed rotor In addition to retrieving saved programs the easy to use front panel can adjust accel/decel rates and change the end of run alarm tone Rotor exchange is simple and requires less than one minute An included rotor wrench can be used to loosen and install all rotors As soon as the rotor is placed on the motor shaft the identification process activates and the installed rotor code is displayed on the backlit LCD display
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.