Learn More
Sino Biological Influenza A H7N7 Hemagglutinin / HA Antibody, Rabbit MAb 500ug
Supplier:  Sino Biological 11082R019500
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza A H7N7 HA / Hemagglutinin protein.
Catalog No. 50-255-4859
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.