Learn More
Sino Biological H7N7 NA PAb 20UL 40202-T62-20
Supplier:  Sino Biological 40202T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N7 (A/Netherlands/219/2003) Neuraminidase / NA ( Catalog#40202-V07H; AAR11367.1; His36-Ser471). Influenza A H7N7 (A/Netherlands/219/2003) Neuraminidase / NA specific IgG was purified by Influenza A H7N7 (A/Netherlands/219/2003) Neuraminidase / NA affinity chromatography.
Catalog No. 50-241-0693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.