Learn More
Abcam HA-1077 (Fasudil), ROCK2 Inhibitor, Solid, 10 mg
Description
- Purity: >98%
- Molecular Weight: 327.8 Da
- Biological Activity: ROCK2 inhibitor (IC50 = 1.9 μM); also inhibits PRK2 (IC50 = 4 μM), MSK1 (IC50 = 5 μM), Rsk2 (IC50 = 15 μM), PKA, p70 S6 Kinase, CaM Kinase and MLCK
- Mechanism: Calcium antagonist and vasodilator
- Form: Solid
- Constituents: 100% Fasudil hydrochloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | ROCK2, PRK2, MSK1, Rsk2, PKA, p70 S6 Kinase, CaM Kinase, MLCK |
| Form | Solid |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% Fasudil hydrochloride |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Formulation | 100% Fasudil hydrochloride |
| Quantity | 10 mg |
| For Use With (Application) | ROCK2 inhibition, PRK2 inhibition, MSK1 inhibition, Rsk2 inhibition, PKA inhibition, p70 S6 Kinase inhibition, CaM Kinase inhibition, MLCK inhibition, Calcium antagonism, Vasodilation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.