Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam HA-1077 (Fasudil), ROCK2 Inhibitor, Solid, 50 mg

Catalog No. 502707698
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7698 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7698 Supplier Abcam Supplier No. AB12030650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent ROCK2 inhibitor that also functions as a calcium antagonist and vasodilator.

  • Molecular Weight: 327.8 Da
  • Purity: >98%
  • Potency: ROCK2 inhibitor (IC50 = 1.9 μM); also inhibits PRK2 (IC50 = 4 μM), MSK1 (IC50 = 5 μM), Rsk2 (IC50 = 15 μM), PKA, p70 S6 Kinase, CaM Kinase and MLCK
  • Mechanism: Calcium antagonist and vasodilator
  • Composition: 100% Fasudil hydrochloride
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule ROCK2, PRK2, MSK1, Rsk2, PKA, p70 S6 Kinase, CaM Kinase, MLCK
Buffer 100% Fasudil hydrochloride
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% Fasudil hydrochloride
Quantity 50 mg
For Use With (Application) ROCK2 inhibition, PRK2 inhibition, MSK1 inhibition, Rsk2 inhibition, PKA inhibition, p70 S6 Kinase inhibition, CaM Kinase inhibition, MLCK inhibition, Vasodilation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.