Learn More
CPC Scientific H-Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific TAGP004A
Encompass
This peptide belongs to the influenza hemagglutinin (HA) family and is responsible for attaching the virus to cell receptors and initiating infection.ONE-LETTER SEQUENCE: YPYDVPDYAMOLECULAR FORMULA: C53H67N9O17MOLECULAR WEIGHT:1102.15STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [92000-76-5], SYNONYMS: Influenza Hemagglutinin (HA) Peptide, Anti-HA, HA tag peptide, HA1 fragment 98-106, Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-AlaRESEARCH AREA: AntimicrobialREFERENCES: Schneider, C. et al. Arch. Virol. 125, 103 (1992); Sleigh, MJ. et al. J. Virol. 37, 845 (1981); Czech, M. et al. J. Cell Biol. 123, 127 (1993).
Catalog No. 50-211-0533
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.