Learn More
Invitrogen™ HABP2 Monoclonal Antibody (OTI5H3)

Description
HABP2 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | HABP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI5H3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | HABP2 |
| Gene Accession No. | Q14520 |
| Gene Alias | AI035669; Factor seven-activating protease; factor VII activating protein; factor VII-activating protease; FSAP; HABP; Habp2; Hepatocyte growth factor activator-like protein; HGFAL; hyaduronic acid-binding protein 2; hyaluronan binding protein 2; hyaluronan-binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronan-binding protein 2 50 kDa heavy chain; Hyaluronan-binding protein 2 50 kDa heavy chain alternate form; hyaluronic acid binding protein 2; NMTC5; PHBP; plasma hyalurona; plasma hyaluronan binding protein; plasma hyaluronan-binding protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.