Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HABP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570038 Supplier Invitrogen™ Supplier No. PA570038
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

HABP2 is an extracellularserine protease that binds hyaluronic acid and is involved in celladhesion. The encoded protein is synthesized as a single chain, butthen undergoes an autoproteolytic event to form the functionalheterodimer. Further autoproteolysis leads to smaller, inactivepeptides. This protease is known to cleave urinary plasminogenactivator, coagulation factor VII, and the alpha and beta chains offibrinogen, but not prothrombin, plasminogen, or the gamma chain offibrinogen. Two transcript variants encoding different isoformshave been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HABP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HABP2
Gene Accession No. Q8K0D2
Gene Alias AI035669; Factor seven-activating protease; factor VII activating protein; factor VII-activating protease; FSAP; HABP; Habp2; Hepatocyte growth factor activator-like protein; HGFAL; hyaduronic acid-binding protein 2; hyaluronan binding protein 2; hyaluronan-binding protein 2; Hyaluronan-binding protein 2 27 kDa light chain; Hyaluronan-binding protein 2 27 kDa light chain alternate form; Hyaluronan-binding protein 2 50 kDa heavy chain; Hyaluronan-binding protein 2 50 kDa heavy chain alternate form; hyaluronic acid binding protein 2; NMTC5; PHBP; plasma hyalurona; plasma hyaluronan binding protein; plasma hyaluronan-binding protein
Gene Symbols HABP2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human HABP2 (aa 379-428).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226243
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.