Learn More
Thermo Scientific™ HAEMAFlex™ 12/16 Swinging Bucket Rotors
GSA_VA
Catalog No. p-6545038
Change view
Click to view available options
Description:
HAEMAFlex 12 Swinging Bucket Rotor
HAEMAFlex 16 Swinging Bucket Rotor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
75-003-862 HAEMAFlex 12 Swinging Bucket Rotor
75-003-882 HAEMAFlex 16 Swinging Bucket Rotor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 75-003-862 Supplier Thermo Scientific™ Supplier No. 75003862
Only null left
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Maximize total blood bags per spin with the unique 16 x 500mL blood bag capacity of the Thermo Scientific™ HAEMAFlex™ 12/16 rotors for the new Thermo Scientific™ blood banking centrifuges.
For high-throughput blood processing needs, the Thermo Scientific™ HAEMAFlex™ 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the Thermo Scientific™ HAEMAFlex™ 16 rotor and the same bucket selections.
The rotors HAEMFlex 12 and 16 can be equipped with adapters to spin the most common blood collection tubes, conical tubes and plates, using oval buckets (750003964).
High-throughput sample processing with the flexibility to match your current and future needs with 12 or 16 x 500mL blood bag (12 or 16 x 1000mL) capacity, utilizing the same bucket system.
These windshielded rotors feature Thermo Scientific™ Eco-Spin™ technology, providing energy savings up to 60%, based on an engineering evaluation of windshielded and non-windshielded designs of the same rotor body.
Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ functions enabling instant rotor lid opening, storage and rotor identification to save time and protects the integrity of your samples.
Rotor buckets with Thermo Scientific™ Dura-Coat™ nickel-coating are also available, providing protection against corrosion due to moisture, chemicals or alkaline solutions that otherwise can weaken the structural integrity of a metal rotor bucket.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description HAEMAFlex 12 Swinging Bucket Rotor
Material Stainless Steel Rotor Body, Aluminum Windshield and Bucket, Polypropylene Bottle
Type Swinging Bucket Rotor
Capacity 12 x 500 mL blood bags (12 x 1000 mL), 312 x 5/7 mL blood tubes (oval buckets 75003964 required)
Max. RCF 7,187 x g
Max. Speed 4,700 rpm
For Use With (Equipment) Thermo Scientific Cryofuge 8 and 16 and Sorvall BP 8 and 16 Centrifuges