Learn More
Haemo-Sol Haemo - Sol Enzyme Active, 5lb.
SDP
Supplier:  Haemo-Sol 026055
Encompass_Preferred
Haemo-Sol Enzyme Active (026-055) is designed for use in situations where items are not cleaned immediately after use or are left to sit for long periods of time. Enzymes in this formulation digest and break down the protein binders that hold proteinaceous wastes and other contaminants to the apparatus, instrument, or part being cleaned. Haemo-Sol Enzyme Active is effective for all soak-and-rinse and ultrasonic cleaning. It is recommended that customers use ounce (14 grams) to 1 gallon (3.79 liters) of water as a mix ratio. This product contains both non-ionic and anionic surfactants.
Catalog No. NC0573588