Learn More
Haemo-Sol™ Medical and Laboratory Instrument Cleaners
SDP
Catalog No. 23721050
Click to view available options
Product Type:
Non-sudsing Cleaner
Regular Cleaner
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-721-050 Non-sudsing Cleaner
23-721-051 Regular Cleaner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-721-050 Supplier Haemo-Sol™ Supplier No. 026051
Add to Cart
Add to Cart
Completely dissolves blood, serum, mucus, sputum, urine, pus, agar, gelatin, and other protein-based soils from medical and laboratory instruments
Keep items lasting longer and looking brand new!
  • Used in laboratories, hospitals, medical, dental, and veterinary offices, as well as crime labs, environmental organizations, and pharmaceutical companies
  • Simply soak items in Haemo-Sol and rinse – no need to scrub
  • Products are 100% soluble and leave no soil or cleaner residue on parts or instruments
  • Specially formulated to not rust metals or cloud glassware
Specifications
Product Type Non-sudsing Cleaner
For Use With (Application) Laboratories, hospitals, medical, dental, and veterinary offices, as well as by crime labs, electronic manufacturing plants, environmental organizations, and pharmaceutical companies
Volume (English) 5 lb.