Learn More
Liofilchem™ Haemophilus Test Agar
SDP
Catalog No. 21403464 Shop All Liofilchem Products
Change view
Click to view available options
Quantity:
20 Plates
Catalog No. Quantity
21-403-464 20 Plates
1 options
Catalog No. 21-403-464 Supplier Liofilchem™ Supplier No. 10080
Add to Cart
Add to Cart
A medium for susceptibility testing of Haemophilus spp, according to CLSI
  • Haemophilus Test Agar is a medium specifically formulated for the susceptibility testing of Haemophilus influenzae as standardized by CLSI.
  • Facilitates accurate measurement of zones of inhibition (areas where bacterial growth is prevented around antibiotic disks)
  • Designed for susceptibility testing of Haemophilus influenzae and other Haemophilus spp., in accordance with standards such as those recommended by the Clinical and Laboratory Standards Institute (CLSI).
Specifications
Manufacturing Origin Italy
Product Type Prepared Microbiology Plate
Sterility Non-sterile
Target Organism Class Haemophilus species, Fastidious, Haemophilus species
Test Type Antimicrobial Susceptibility
Quantity 20 Plates
Diameter (Metric) 90 mm