Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium AA Standard, 1000 ppm Hf in 3% HNO3/tr HF, Ricca Chemical
SDP

Catalog No. AHF1KN100 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AHF1KN100 100 mL Natural LDPE Bottle
AHF1KN500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AHF1KN100 Supplier Ricca Chemical Company Supplier No. AHF1KN100
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12055-23-1 , 7697-37-2 , 7664-39-3 , 7732-18-5
Molecular Formula HfO2
Molecular Weight (g/mol) 210.49
MDL Number MFCD00003565
InChI Key WIHZLLGSGQNAGK-UHFFFAOYSA-N
IUPAC Name hafnium(4+) dioxidandiide
SMILES [O--].[O--].[Hf+4]

Specifications

Packaging Natural LDPE Bottle
CAS Min % 93.0, 4.49, 0.11, 0.09
CAS Max % 97.0, 4.67, 0.11, 0.1
Odor Odorless
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Concentration or Composition (by Analyte or Components) 1mL 1mg Hf (1, 000ppm Hf) Hf in 3% HNO3 / tr HF
Chemical Name or Material Hafnium AA Standard
Show More Show Less
ShelfLife 24 Months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.