Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hafnium silicide, Thermo Scientific Chemicals

Catalog No. p-7049042
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1256409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1256409 Supplier Thermo Scientific Chemicals Supplier No. 01256409
Only null left
Add to Cart

They have been used as ohmic contacts, Schottky barrier contacts, gate electrodes, local interconnects, and diffusion barriers. FeSi2 has become the remarkable source as high temperature thermoelectric material. It is also used as a powder in ceramic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12401-56-8
Molecular Formula HfSi2
Molecular Weight (g/mol) 234.66
MDL Number MFCD00167026
InChI Key TWRSDLOICOIGRH-UHFFFAOYSA-N
Synonym hafnium silicide
PubChem CID 11230071
IUPAC Name hafnium;silicon
SMILES [Si].[Si].[Hf]

Specifications

Melting Point 1680°C
Physical Form Powder
Quantity 10 g
Solubility Information it is insoluble in water.
Formula Weight 234.66
Odor Odorless
Chemical Name or Material Hafnium silicide
EINECSNumber 235-640-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.