Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hair Cortex Cytokeratin Monoclonal Antibody (AE13)
GREENER_CHOICE

Catalog No. 01674884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-884 Supplier Invitrogen™ Supplier No. MA15765
  • $747.00
    $622.10 / Each of 1

    Save $124.90

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-μ65 detects Cytokeratin, AE13 in human and mouse samples. MA1-μ65 has been successfully used in Western blot, immunocytochemistry, immunofluorescence and immunohistochemistry procedures. The MA1-μ65 immunogen is human hair keratins. This antibody reacts with acidic 44-46 kDa hair cortex keratins.

Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. PCK-26 facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hair Cortex Cytokeratin
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone AE13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene KRT40
Gene Accession No. Q6A162, Q6IFX3
Gene Alias CK-40; cytokeratin-40; K40; Ka36; keratin 40; keratin 40, type I; keratin, type I cytoskeletal 40; Keratin-40; Krt40; ortholog of type I hair keratin KA36; type I hair keratin KA36
Gene Symbols KRT40
Host Species Mouse
Immunogen Human hair keratins from P3X63Ag. 8.653 cells.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125115, 406221
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.