Learn More
TerraCycle™ Hair Nets and Beard Nets Zero Waste Box™



Description
- WHAT YOU CAN RECYCLE
- Please send us any brand and size of used hair and beard nets.
- Do not send in gloves, lab coats, or garments.
- How It Works
- Order: Select a Zero Waste Box system and place your order.
- Collect: Collect the accepted items to fill the system.
- Ship: Send it back to TerraCycle with the prepaid return label on your system.*
- Reorder: Order your next Zero Waste Box system to continue to recycle everything.
- What’s Included?
- One Zero Waste Box
- One prepaid return label, affixed to your box
- Processing and recycling of the materials
- Customer support from our team of experts
- Important Shipping Restrictions
- For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:
- Only send in items this system accepts. Sending in items that fall outside the selected category causes contamination and the need for added separation, which increases costs.
- Do not send in household items that may be considered hazardous waste. These include items such as lithium-ion batteries, pressurized canisters, pesticides, oil-based paint or medical sharps, organics, broken glass, biomedical waste, soiled diapers, or expired medication. Do not send in anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
- Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- Do not ship dripping packages. Please make sure all excess product has been removed before putting it in your Zero Waste Box, Pouch, or Pallet. If you choose to rinse your items, please note that the system must be completely dry prior to shipping.
- For the Zero Waste Box system to work effectively and safely, please follow these simple shipping guidelines on how and what you can ship back to TerraCycle:
Specifications
Specifications
| Capacity (Metric) | 79.31 L |
| Capacity (English) | 79.31 L |
| Color | Brown, Green, White |
| Dimensions (L x W x H) | 11 x 11 x 40 in. |
| Disposable | Single-use |
| Latex Free | Yes |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Includes | Cardboard Box |
| Liner Material | Plastic |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.