Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hairless Monoclonal Antibody (OTI1D9)
GREENER_CHOICE

Catalog No. PIMA526821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526821 Supplier Invitrogen™ Supplier No. MA526821
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hairless Monoclonal Antibody for Western Blot, IHC (P)

Drosophila genes can be categorized based on the type of protein for which they encode and are represented by six major classifications, which include intracellular signaling proteins, transmembrane proteins, RNA binding proteins, secreted factors, transcription regulators (basic helix-loop-helix, homeodomain containing, zinc finger containing and chromatin associated) or other functional proteins. Among these numerous proteins, Hairless (CG5460, CT36411, CT17282) is a transcriptional co-repressor that contains a Paired box domain, a KX-repeat domain, a NEDL-repeat domain and a serine-rich domain. Hairless is an antagonist of the Notch signaling pathway in Drosophila.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hairless
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HR
Gene Accession No. O43593
Gene Alias ALUNC; AU; ba; bldy; HAIR; hair growth associated; hairless; hairless homolog; Hairless protein; hr; HSA277165; HYPT4; Lysine-specific demethylase hairless; MUHH; MUHH1; N; protein hairless; rh; rh-bmh; rhino
Gene Symbols HR
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 946-1189 of HR produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55806
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.