Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Half Life Analogue Demonstration
SCI_ED

Catalog No. S15101 Shop All Eisco Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S15101 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15101 Supplier Eisco™ Supplier No. HALFLIFE
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Half Life Analogue Demonstration allows students to simulate radioactive decay, while also creating a decay curve.

  • An effective hands on demonstration of radioactive decay
  • Allows students to to safely study radioactive particles, while creating a decay curve
  • Set includes 10 cups and 500 cubes (representing nuclei)
  • Each cube has one face identified, when this face is the uppermost face the cube represents a decayed radioactive nuclei
  • Ten groups shake their set of nuclei and roll them onto a table
  • For each throw, the decayed nuclei are counted and removed from the set

Specifications

Grade 7 to 14
Age 12 to 20
For Use With (Application) Studying Radioactive Particles
Includes 10 Cups, 500 Cubes
Class Size 10 Lab Groups
Product Type Half Life Demonstration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.