Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Half Size Youth Torso Anatomical Model with Head
SCI_ED

Catalog No. S012567 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H): 20 x 10.3 x 7.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S012567 Dimensions (L x W x H): 20 x 10.3 x 7.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012567 Supplier Eisco™ Supplier No. AM0015
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This model, representing a young human, is half-life size with 9 removable parts. The head is dissected in half to show the skull and brain, as well as the internal structures of the mouth, neck and sinuses. Bisected lungs, 2 part heart, liver, stomach, small and large intestine and half of right kidney can be removed for closer study.

  • Half size
  • Youth Model
  • 9 parts
  • Removable organs

Specifications

Color Tan, Red
Dimensions (L x W x H) 20 x 10.3 x 7.5 in.
Material Plastic
Weight (English) 5.2 lb.
Gender Male, Female
Product Type Torso Model
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.