Learn More
Medchemexpress LLC 5-[(5-nitro-1,3-thiazol-2-yl)sulfanyl]-1,3,4-thiadiazol-2-amine | 40045-50-9 | MFCD01927019 | 98.9% | 261.30 g/mol | C5H3N5O2S3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10759750MG
Encompass_Preferred
Halicin is a small-molecule JNK inhibitor intended for laboratory research. It exhibits biochemical activity against JNK (IC50 0.7 μM) and disrupts the JNK-JIP protein-protein interaction (IC50 239 nM). The compound is provided as a solid and as a DMSO solution for flexible assay preparation.
Catalog No. 50-002-40184
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.