Learn More
United Scientific™ Hall's Carriage
SDPSCI_ED
Catalog No. S00077 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Hall's Carriage
Catalog No.
S00077 Hall's Carriage
1 options
Catalog No. S00077 Supplier United Scientific™ Supplier No. HLCRG1E
Add to Cart
Add to Cart
United Scientific Hall's Carriage is perfect for demonstrations and investigating principles such as Newton’s laws, acceleration, velocity and friction.
This economy Hall’s carriage is perfect for demonstrations and investigating principles such as Newton’s laws, acceleration, velocity and friction.
  • This economy Hall’s carriage has a durable plastic body with a roomy well for additional masses (not included).
  • The hole on the front bumper allows you to easily connect a string for pulling the cart up a ramp.
  • Both the body and wheels are made from high impact plastic.
  • Dimensions of the carriage are 5.25 in. L x 2.25 in. W x 1.5 in. H and the well size is 2 in. L x 2.25 in. W x 0.95 in. H.
Specifications
Product Type Hall's Carriage