Learn More
United Scientific Supplies Hall's Carriage with Universal Smartphone Adapter
SDPSCI_ED
Catalog No. S27813 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Includes:
Hall's Carraige, Smartphone Holder, Activity Guide
Catalog No. Includes
S27813 Hall's Carraige, Smartphone Holder, Activity Guide
1 options
Catalog No. S27813 Supplier United Scientific Supplies Supplier No. HLCRG1USA
Add to Cart
Add to Cart
Utilize the power of a smartphone with this unique Hall’s Carriage
  • Hall's carriage securely holds a smartphone for motion experiments
  • Graph acceleration and velocity with preferred app
  • Includes Activity Guide
Specifications
Product Type Hall's Carraige Motion Experiment
For Use With (Application) Motion Experiment
Includes Hall's Carraige, Smartphone Holder, Activity Guide