Learn More
Advanced Materials Technology HALO 1000 DIPHENYL 2.7
Supplier:  Advanced Materials Technology 92710726
Encompass_Preferred
HALO BIOCLASS Diphenyl 1000 Å Protein columns made with innovative large pore Fused-Core particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns. HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Catalog No. 50-255-6445
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.