Learn More
Advanced Materials Technology HALO 120 A OLIGO C18 2.7 um 4.6 x 150 mm
Supplier:  Advanced Materials Technology P2A64702
Encompass_Preferred
The HALO OLIGO C18 incorporates surface modified organo-silane technology for alkaline resistance, resulting in excellent stability under elevated pH operating conditions common in oligonucleotide separation methods.  The inert column hardware also addresses absorptivity concerns so the highest recoveries can be achieved.
Catalog No. NC3596292
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.