Learn More
Advanced Materials Technology HALO 400 C4, 3.4, 2.1 x 50 mm
Supplier:  Advanced Materials Technology 93412414
Encompass_Preferred
HALO 400 C4, 3.4, 2.1 x 50 mm
Catalog No. 50-274-5760
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.