Learn More
Advanced Materials Technology HALO ES-C18 2UM 2.1X5 3PK
Supplier:  Advanced Materials Technology 91122102
Encompass_Preferred
HALO BIOCLASS ES-C18 Peptide columns made with innovative Fused-Core particle technology are ideal for both ultrafast and ultrahigh resolution separations of peptides and polypeptides up to 20 kDa. HALO peptide columns offer comparable peak capacity to sub-2 ?m non-core columns at 40¿50% back pressure (2.7 ?m), and 20% higher peak capacity than sub-2 ?m non-core columns at comparable back pressure (2 ?m). HALO 160 ¿peptide columns are optimized for high performance in HPLC, UHPLC, and LCMS peptide applications.
Catalog No. NC2459962
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.