Learn More
CleanSpace™ HALO Particulate Filter HEPA Low Profile (Pk3)
Suitable for protection against airborne particulates (dust, mists and fumes) and biohazards
Supplier: CleanSpace™ CS3022
Description
- Used with the revolutionary CleanSpace™ PAPR: light weight, no hoses or belts
- Suitable for protection against airborne particulate (dust, mists and fumes).
- Materials: Fibreglass particulate media and plastic casing, silicone seal
- Easy and quickly fitted and removed from the power unit
- Pack of 3 (CS3002)
Specifications
Filter | |
CS3020 | |
3 Single HEPA Filter |