Learn More
Ushio America, Inc. Tungsten Halogen Lamp Bulb, 150W 21V
Supplier: Ushio America, Inc. 1000297
This item has a minimum qty of 10 per supplier requirements.
Tungsten Halogen Lamp, Overall Length: 44.5mm, Reflector Type: Specular, Filament: CC-6, Working Distance: 28mm, Color Temperature: 3400 deg.K, Avg. Life: 40 hr., Burn Position: BD/Hor, 2-Pin Base GX5.3 MR-16 Reflector
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.