Learn More
World Precision Instrument Halogen Replacement Lamp for D4H
SDP
Catalog No. 502539429
Encompass
  • Replacement halogen lamp for D4H
  • 360-2500 nm
  • 900 hrs
  • 3100K
Catalog No. 50-253-9429 Supplier World Precision Instrument Supplier No. 503848
  • Replacement halogen lamp for D4H
  • 360-2500 nm
  • 900 hrs
  • 3100K