Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Haloperidol USP
SDP

Catalog No. 14186457 Shop All Medisca Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-457 25 g
14-185-132 100 g
14-186-455 5 g
14-185-462 10 g
14-185-466 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-186-457 Supplier Medisca Supplier No. 033004
Add to Cart
Add to Cart

Haloperidol USP is a white to faintly yellowish, amorphous or microcrystalline powder.

Haloperidol USP is a white to faintly yellowish, amorphous or microcrystalline powder.

Specifications

Chemical Name or Material Haloperidol USP
Packaging Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT
CAS 52-86-8
Type Drug Standard
Molecular Weight (g/mol) 375.8687
Quantity 25 g
Percent Purity 98 to 102 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.