Learn More
Halyard Health Universal Precautions Lab Coats
Catalog No. 23500171
Change view
Click to view available options
Color:
Blue
White
Size:
Small
Medium
Large
X-Large
2X-Large
Length (Metric):
105.4 cm
116.8 cm
118.1 cm
119.4 cm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Length (Metric)
23-500-171 White Large 118.1 cm
23-500-169 White Small 105.4 cm
23-500-170 White Medium 116.8 cm
23-500-172 White X-Large 118.1 cm
23-500-173 White 2X-Large 119.4 cm
23-500-174 Blue Small 105.4 cm
23-500-175 Blue Medium 116.8 cm
23-500-176 Blue Large 118.1 cm
23-500-177 Blue X-Large 118.1 cm
23-500-178 Blue 2X-Large 119.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 23-500-171 Supplier Halyard Health Supplier No. 10042
Add to Cart
Add to Cart
This item is not returnable. View return policy
Protective tri-layer fabric offers resistance to blood strikethrough at various levels of exposure
  • For low to moderate exposure to fluids or chemicals
  • Traditional collar and knit cuffs
  • Six snaps
  • Three reinforced pockets
  • Back vent
  • Neck loop for hanging
  • Size/name label
  • Available in colors blue and white
Specifications
Size Large
Material SMS Fabric
Color White
Cuff Style Knit
Description 3-layer SMS fabric
Size (Metric) Chest 67.75 cm
Length (Metric) 118.1 cm