Learn More
Hamilton™ 1000 Series Syringe, Without Needle, Sold by MilliporeSigma™ Supelco™
Catalog No. 111016260 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
1 mL
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Connection Type:
Kel-F Luer Lock with Slots
Luer Lock with Slots
Luer Tip, Kel-F Hub
Metal (N) or Kel-F
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Connection Type
11-101-6260 5 mL Luer Lock with Slots
11-100-9672 25 mL Kel-F Luer Lock with Slots
11-100-9346 50 mL Kel-F Luer Lock with Slots
11-101-0882 2.5 mL Luer Lock with Slots
11-101-0845 10 mL Luer Lock with Slots
11-101-1422 1 mL Metal (N) or Kel-F
11-101-1111 2.5 mL Metal (N) or Kel-F
11-101-1181 10 mL Metal (N) or Kel-F
11-100-9791 25 mL Metal (N) or Kel-F
11-100-9485 50 mL Metal (N) or Kel-F
11-100-9031 100 mL Metal (N) or Kel-F
11-101-1203 1 mL Luer Tip, Kel-F Hub
11-101-1285 2.5 mL Luer Tip, Kel-F Hub
11-101-1026 5 mL Luer Tip, Kel-F Hub
11-101-0836 10 mL Luer Tip, Kel-F Hub
11-101-0680 1 mL Luer Lock with Slots
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
Catalog No. 11-101-6260 Supplier MilliporeSigma Supelco Supplier No. 26210U
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Gas-tight design minimizes the gas exchange in GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed. Tip is threaded in a luer lock syringe which allows the needle to be tightly screwed to the tip.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Gastight™ Syringe
Model 1005 TLL
Quantity 1 Ea.
Volume (Metric) 5 mL
Product Line Hamilton
For Use With (Equipment) Hamilton 1005 TLL
Connection Type Luer Lock with Slots
GASTIGHT is a registered trademark of Hamilton Co.