Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Hamilton™ 700 Series Syringe, Cemented Needle (N), Sold by MilliporeSigma™ Supelco™

Catalog No. 111011170 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Gauge:
23s G
26s G
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge
11-101-1170 23s G
11-101-0351 26s G
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1170 Supplier MilliporeSigma™ Supelco™ Supplier No. 26228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides accurate volume delivery with a plunger that is mostly unsurpassed

TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel (Tip)
Description 700 Series
Needle Gauge 23s G
Length (English) Needle 1.71 in.
Product Type Syringe
Needle Point Number 1
Quantity 1 Ea.
Volume (Metric) 10 μL
Product Line Hamilton
Length (Metric) Needle 43 mm
Needle Point Style Cone
For Use With (Equipment) Merlin Microseal
Connection Type Cemented
Show More Show Less

Hamilton is a registered trademark of Hamilton Co.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.