Learn More
Hamilton™ LC Syringe, Sold by MilliporeSigma™ Supelco™
Catalog No. 111010046 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Volume (Metric):
25 μL
Catalog No. Volume (Metric)
11-101-0046 25 μL
1 options
Catalog No. 11-101-0046 Supplier MilliporeSigma Supelco Supplier No. 29601U
Add to Cart
Add to Cart
For use with Waters WISP™
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (Barrel), PTFE (Plunger)
Model 1702
Quantity 1 Ea.
Volume (Metric) 25 μL
For Use With (Equipment) Waters WISP HPLC Autosamplers
Waters is a registered trademark of Waters