Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamilton™ Needles, 2 in., Sold by MilliporeSigma™ Supelco™

Catalog No. 111011625 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Point Style:
Bevel
Blunt
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Point Style
11-101-1625 Blunt
11-101-1595 Bevel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-1625 Supplier MilliporeSigma Supelco Supplier No. 19143U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Removable Needle (RN) Syringes

TRUSTED_SUSTAINABILITY

Specifications

Material Stainless Steel (Tip)
Needle Gauge 22 G
Length (English) Needle 2 in.
Product Type Needle
Needle Point Number 3
Quantity 6 Pk.
Volume (Metric) 250 to 10,000 μL
Length (Metric) Needle 51 mm
Needle Point Style Blunt
For Use With (Equipment) Removable Needle Syringe

Hamilton is a registered trademark of Hamilton Co.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.