Learn More
Sigma Aldrich Fine Chemicals Biosciences Atto 647N DOPE suitable fo1MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 42247
Encompass
Atto 647N belongs to a new generation of fluorescent labels for the red spectral region. The dye is designed for application in the area of life science e.g. labeling of DNA RNA or proteins. Characteristic features of the label are strong absorption excellent fluorescence quantum yield high photostability excellent ozone resistance good solubility and very little triplet formation. Atto 647N is a cationic dye. After coupling to a substrate the dye carries a net electrical charge of 1. In common with most Atto-labels absorption and fluorescence are independent of pH in the range of 2 to 11 used in typical applications. As supplied Atto 647N consists of a mixture of two isomers with practically identical absorption and fluorescence properties.Atto-Dye Labeled Phospholipids Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head gro
Catalog No. 50-290-9051
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.