Learn More
Sigma Aldrich Fine Chemicals Biosciences Peristaltic pump 8 Channel1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences Z741761
IBI offers a high quality 8-channel peristaltic pump for use with the IBI flow cell systems. The 8-channel pump can accommodate two each of the 3-channel flow cells or up to four each of the convertible flow cells.The pump has a unique planetary gear drive system with 8 rollers that provides the most gentle fluid delivery of any multi-channel peristaltic pump which is ideal for use with the IBI flow cells systems. The pump utilizes stoppered tubing that accommodates the cassette design of the pump. IBI flow cell assemblies come complete with the necessary pump tubing already in place. (Except cell ONLY models).