Learn More
Hamilton™ Removable Needle to Luer Adapters
Catalog No. 14819284 Shop All Hamilton Company Products
Change view
Click to view available options
Packaging Quantity:
1/Pk.
Catalog No. Packaging Quantity
14-819-284 1/Pk.
1 options
Catalog No. 14-819-284 Supplier Hamilton™ Supplier No. 35083
Add to Cart
Add to Cart
These adapters convert a Hamilton specific termination to an industry standard luer connection.
  • Compatible with SampleLock or Removable Needle syringes 250μL and larger
  • Internal diameter is 1mm making it possible to spike samples with an internal standard using a 26 gauge needle or smaller
  • Male luer lock connection
Specifications
Type Male Luer-lok Hub Adapter, for 10 to 33 ga. Needles
Packaging Quantity 1/Pk.
Port Type Male Luer Lock
For Use With Removable Needle Syringe (250μL and Larger)