Learn More
Hamilton™ Syringe Guide for Microliter™ and Gaslight™ Syringes
Catalog No. 1481930 Shop All Hamilton Company Products
Change view
Click to view available options
For Use With (Equipment):
Microliter, Gastight and 700 and 1700 Series Syringes
Microliter, Gastight, 7000 Series and 25 to 500μL Syringes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14-819-30 Microliter, Gastight, 7000 Series and 25 to 500μL Syringes
14-819-20 Microliter, Gastight and 700 and 1700 Series Syringes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-819-30 Supplier Hamilton™ Supplier No. 14906
Add to Cart
Add to Cart
Designed to work with 700/7000 series MICROLITER syringes and 1700 series GASTIGHT syringes
Syringe guides are designed to prevent plunger bending and the adjustable stop prevents plunger damage.
Specifications
Product Type Syringe Guide
For Use With (Equipment) Microliter, Gastight, 7000 Series and 25 to 500μL Syringes