Learn More
MilliporeSigma™ Supelco™ Hamilton™ Valve Flushing Syringe, 1001
Catalog No. 111010873 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Needle Point Style:
3
Catalog No. Needle Point Style
11-101-0873 3
1 options
Catalog No. 11-101-0873 Supplier MilliporeSigma™ Supelco™ Supplier No. 58643
 Restricted Item
Specifications
Material Stainless Steel (Tip)
Product Type Blunt Tip
Volume (Metric) 1 mL
Needle Point Style 3
For Use With (Equipment) Rheodyne, SSI, VISF-2
Hamilton is a registered trademark of Hamilton Co.