Learn More
Hamilton™ VDRL Bent Needle
Catalog No. 14816230 Shop All Hamilton Company Products
Change view
Click to view available options
Needle Gauge:
18 G
Catalog No. Needle Gauge
14-816-230 18 G
1 options
This item is not returnable. View return policy
Catalog No. 14-816-230 Supplier Hamilton™ Supplier No. 90432
Add to Cart
Add to Cart
This item is not returnable. View return policy
Needle used for VDRL Antigen Test, a microscopic nontreponemal flocculation test to be used for the detection of reagin
Specifications
Autoclavable Autoclavable
Needle Gauge 18 G
Length (English) Needle 2.65 in.
Product Type Replacement Syringe Needle
Needle Point Style Square/Blunt