Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HAMP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542038 Supplier Invitrogen™ Supplier No. PA542038
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MALSSQIWAA CLLLLLLLAS LTSGSVFPQQ TGQLAELQPQ DRAGARASWM Sequence homology: Human: 100%; Rat: 83%.

HAMP is involved in the maintenance of iron homeostasis, and it is necessary for the regulation of iron storage in macrophages, and for intestinal iron absorption. The preproprotein is post-translationally cleaved into mature peptides of 20, 22 and 25 amino acids, and these active peptides are rich in cysteines, which form intramolecular bonds that stabilize their beta-sheet structures. These peptides exhibit antimicrobial activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HAMP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HAMP
Gene Accession No. P81172
Gene Alias HAMP; Hamp1; HEPC; Hepc1; Hepc20; Hepc25; Hepcidin; hepcidin antimicrobial peptide; hepcidin antimicrobial peptide 1; Hepcidin-20; Hepcidin-25; HFE2B; LEAP1; LEAP-1; liver-expressed antimicrobial peptide 1; PLTR; Putative liver tumor regressor; UNQ487/PRO1003
Gene Symbols HAMP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HAMP (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57817
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.