Learn More
ILEX LIFE SCIENCES LLC HAMS F12 P04-14559 500 ML
Supplier: ILEX LIFE SCIENCES LLC P0414559
PAN-Biotech Ham's F12 Medium, w/o: L-Glutamine, w/o: Phenol red, w: 1.176 g/L NaHCO3 (500 ml). BICARBONATE: With 1.176 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: Without glutamine. PHENOL RED: Without Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Ham's F12 nutrient mix was developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells, and mouse L-cells. It can be used for serum-free growth of CHO cultures supplemented with hormones, selenium and other supplements as well as for other mammalian cells, especially cell lines of rodent origin (particularly rabbit and rat).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.