Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hamster anti-IgG1, κ, Isotype Control, APC-R700, Clone: A19-3, BD

Catalog No. BDB565129 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565129 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565129 Supplier BD Biosciences Supplier No. 565129
Add to Cart
Add to Cart

Specifications

Antigen IgG1, κ
Applications Flow Cytometry
Classification Monoclonal
Clone A19-3
Conjugate APC-R700
Formulation Aqueous buffered solution containing BSA, protein stabilizer, glycerol, and ≤0.09% sodium azide.
Host Species Hamster
Immunogen TNP-keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Content And Storage Store undiluted at 4°C. Do not freeze. Protect from light.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.