Learn More
MyBioSource Hamster Carboxylesterase 1 ELISA Kits, Carboxylesterase 1
SDP
Supplier:  MyBioSource MBS086978
Encompass_Preferred
The MyBioSource Hamster Carboxylesterase 1 ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of hamster carboxylesterase 1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. The kit has a detection range of 0.156-10 ng/mL and includes a 96-well strip plate pre-coated with an antibody specific for carboxylesterase 1, standards, detection reagents A and B, wash and stop solutions, and an instruction manual. The total assay time is around 3.5 hours. This research-use-only ELISA kit can detect hamster carboxylesterase 1 with high sensitivity.
Catalog No. NC2831175
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.