Learn More
MyBioSource Endothelial Lipase
SDP
Supplier:  MyBioSource MBS005633
Encompass_Preferred
The product is human endothelial lipase protein, recombinant, expressed in HEK 293 cells. It is supplied as a lyophilized powder containing 0.05M Tris, 0.15M NaCl, pH 7.5. The molecular weight is approximately 68 kDa. This product can be used for applications such as ELISA, Western Blot, and cell culture. When reconstituted with sterile water, the concentration is 0.1 mg/mL. The product should be stored at -20°C.
Catalog No. NC2831152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.